ENA156508504 Enamine

8-(chloromethyl)-1,3-dimethyl-2,3,6,9-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD09971380
  • Cas Number : 64398-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31037
Availability details for
Quantity
  • Availability
  • Add to Cart