ENA156508535 Enamine

1-(3-fluorobenzoyl)-1,4-diazepane

  • MDL number MFCD09043044
  • Cas Number : 926215-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31049
Availability details for
Quantity
  • Availability
  • Add to Cart