ENA156508537 Enamine

5-propyl-5H-[1,2,4]triazino[5,6-b]indole-3-thiol

  • MDL number MFCD01306247
  • Cas Number : 36047-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31052

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31052
Availability details for
Quantity
  • Availability
  • Add to Cart