ENA156508552 Enamine

3-(1,3,4-oxadiazol-2-yl)aniline

  • MDL number MFCD00099501
  • Cas Number : 5378-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31065
Availability details for
Quantity
  • Availability
  • Add to Cart