ENA156508556 Enamine

2-(4-chlorophenyl)pyrrolidine

  • MDL number MFCD01631838
  • Cas Number : 38944-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31069

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-31069
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart