ENA156508557 Enamine

3-[2-(trifluoromethyl)phenyl]-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD09971384
  • Cas Number : 1038770-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31070
Availability details for
Quantity
  • Availability
  • Add to Cart