ENA156508561 Enamine

(6-pyrrolidin-1-ylpyridin-3-yl)methylamine

  • MDL number MFCD09049100
  • Cas Number : 754977-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31075

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31075
Availability details for
Quantity
  • Availability
  • Add to Cart