ENA156508565 Enamine

6-bromoquinazoline-4-thiol

  • MDL number MFCD09042588
  • Cas Number : 544461-20-3
  • Molecular Weight : 241.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31076
Availability details for
Quantity
  • Availability
  • Add to Cart