ENA156508570 Enamine

2-chloro-N-(chloroacetyl)-N-(3,4,5-trimethoxyphenyl)acetamide

  • MDL number MFCD09971387
  • Cas Number : 1000932-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31080

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31080
Availability details for
Quantity
  • Availability
  • Add to Cart