ENA156508580 Enamine

1-sulfamoylpiperidine-4-carboxylic acid

  • MDL number MFCD09940510
  • Cas Number : 1000932-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31102
Availability details for
Quantity
  • Availability
  • Add to Cart