ENA156508581 Enamine

1-sulfamoylpiperidin-4-yl 2,2,2-trifluoroacetate

  • MDL number MFCD09971394
  • Cas Number : 1000932-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31103
Availability details for
Quantity
  • Availability
  • Add to Cart