ENA156508582 Enamine

4-benzylpiperazine-1-sulfonamide

  • MDL number MFCD09971395
  • Cas Number : 178312-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31104
Availability details for
Quantity
  • Availability
  • Add to Cart