ENA156508587 Enamine

methyl 4-[(carbamimidoylsulfanyl)methyl]benzoate hydrobromide

  • MDL number MFCD02130300
  • Cas Number : 479580-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Product Link https://www.enaminestore.com/catalog/EN300-31110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31110
Availability details for
Quantity
  • Availability
  • Add to Cart