ENA159843315 Enamine

4-(4-fluorophenoxy)butan-1-amine hydrochloride

  • MDL number MFCD08273543
  • Cas Number : 1171603-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15235

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15235
Availability details for
Quantity
  • Availability
  • Add to Cart