ENA159843317 Enamine

2-chloro-1-[4-(3-nitrobenzenesulfonyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD09971328
  • Cas Number : 1050909-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23035
Availability details for
Quantity
  • Availability
  • Add to Cart