ENA159843322 Enamine

2-chloro-N-[(4-fluoro-3-methylphenyl)methyl]acetamide

  • MDL number MFCD09971338
  • Cas Number : 1050909-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28336
Availability details for
Quantity
  • Availability
  • Add to Cart