ENA159843329 Enamine

8-chloro-3,4-dihydro-2H-1,5-benzodioxepin-7-amine hydrochloride

  • MDL number MFCD09971354
  • Cas Number : 1171401-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30410
Availability details for
Quantity
  • Availability
  • Add to Cart