ENA159843338 Enamine

N-(2,2,2-trifluoroethyl)-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD09971424
  • Cas Number : 1078162-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31216
Availability details for
Quantity
  • Availability
  • Add to Cart