ENA159843339 Enamine

5-bromo-2-(4-methylpiperazin-1-yl)aniline dihydrochloride

  • MDL number MFCD09971443
  • Cas Number : 1172842-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31272

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-31272
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart