ENA159843341 Enamine

4-(aminomethyl)-N-methylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD09971463
  • Cas Number : 116599-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31336
Availability details for
Quantity
  • Availability
  • Add to Cart