ENA159843345 Enamine

4-[(difluoromethyl)thio]aniline hydrochloride

  • MDL number MFCD09971488
  • Cas Number : 477593-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222...224
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31427

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31427
Availability details for
Quantity
  • Availability
  • Add to Cart