ENA159843354 Enamine

5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD09971529
  • Cas Number : 69146-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31647
Availability details for
Quantity
  • Availability
  • Add to Cart