ENA159843355 Enamine

[5-(pyrrolidine-1-sulfonyl)thiophen-2-yl]methanamine hydrochloride

  • MDL number MFCD09971536
  • Cas Number : 1170959-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31678
Availability details for
Quantity
  • Availability
  • Add to Cart