ENA159843356 Enamine

N-(4-bromophenyl)-2-(ethylamino)acetamide hydrochloride

  • MDL number MFCD09971537
  • Cas Number : 1171588-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31680
Availability details for
Quantity
  • Availability
  • Add to Cart