ENA159843358 Enamine

2,3,4,9-tetrahydro-1H-carbazol-6-amine hydrochloride

  • MDL number MFCD01739721
  • Cas Number : 1079-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31709
Availability details for
Quantity
  • Availability
  • Add to Cart