ENA159843362 Enamine

5-(1-chloroethyl)-3-(thiophen-2-yl)-1,2,4-oxadiazole

  • MDL number MFCD09971553
  • Cas Number : 1039967-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-31743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31743
Availability details for
Quantity
  • Availability
  • Add to Cart