ENA159843373 Enamine

1-(4-nitrophenyl)-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD09971577
  • Cas Number : 65973-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31875

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31875
Availability details for
Quantity
  • Availability
  • Add to Cart