ENA159843377 Enamine

1-(4-fluorophenyl)-2-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD09971581
  • Cas Number : 1019058-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31899
Availability details for
Quantity
  • Availability
  • Add to Cart