ENA159843381 Enamine

N-[(pyridin-3-yl)methyl]piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD09971587
  • Cas Number : 1170206-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31917
Availability details for
Quantity
  • Availability
  • Add to Cart