ENA159843390 Enamine

1-[2-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD09971602
  • Cas Number : 1049872-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31997
Availability details for
Quantity
  • Availability
  • Add to Cart