ENA159843391 Enamine

1-(4-aminophenyl)-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD09971603
  • Cas Number : 1050884-19-9
  • Molecular Weight : 227.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32008
Availability details for
Quantity
  • Availability
  • Add to Cart