ENA159843400 Enamine

1-{[5-(2-nitrophenyl)thiophen-2-yl]methyl}piperazine

  • MDL number MFCD09971616
  • Cas Number : 1050910-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33306
Availability details for
Quantity
  • Availability
  • Add to Cart