ENA159843402 Enamine

1-(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)piperazine hydrochloride

  • MDL number MFCD09971618
  • Cas Number : 1171471-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33309
Availability details for
Quantity
  • Availability
  • Add to Cart