ENA159843404 Enamine

N-(1-{5-[(ethylamino)methyl]-1,2,4-oxadiazol-3-yl}cyclopentyl)acetamide hydrochloride

  • MDL number MFCD09971620
  • Cas Number : 1171023-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33319
Availability details for
Quantity
  • Availability
  • Add to Cart