ENA159843408 Enamine

3-methyl-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD09971626
  • Cas Number : 1049605-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33350
Availability details for
Quantity
  • Availability
  • Add to Cart