ENA159843409 Enamine

4-[5-(aminomethyl)furan-2-yl]benzene-1-sulfonamide hydrochloride

  • MDL number MFCD09971627
  • Cas Number : 1172734-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33351
Availability details for
Quantity
  • Availability
  • Add to Cart