ENA159843410 Enamine

N-[3-(aminomethyl)phenyl]pyridine-3-carboxamide dihydrochloride

  • MDL number MFCD09971628
  • Cas Number : 1170409-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33352
Availability details for
Quantity
  • Availability
  • Add to Cart