ENA159843416 Enamine

N-(2,2,2-trifluoroethyl)pyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD09971635
  • Cas Number : 1078163-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33375
Availability details for
Quantity
  • Availability
  • Add to Cart