ENA159843421 Enamine

3-phenyl-5H,6H,7H,8H,9H-imidazo[1,5-a]azepine-1-carboxylic acid

  • MDL number MFCD09971640
  • Cas Number : 109650-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33397
Availability details for
Quantity
  • Availability
  • Add to Cart