ENA159843425 Enamine

methyl 5-(chloromethyl)-3-methyl-2-furoate

  • MDL number MFCD09971644
  • Cas Number : 34878-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33423

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-33423
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart