ENA159843426 Enamine

3-chloro-4-(2-methylpropoxy)aniline hydrochloride

  • MDL number MFCD09971646
  • Cas Number : 1170938-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33429
Availability details for
Quantity
  • Availability
  • Add to Cart