ENA159843429 Enamine

N-benzylpiperazine-1-sulfonamide hydrochloride

  • MDL number MFCD09971649
  • Cas Number : 1171172-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33436
Availability details for
Quantity
  • Availability
  • Add to Cart