ENA159843432 Enamine

2-amino-N-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-5-yl)acetamide hydrochloride

  • MDL number MFCD09971654
  • Cas Number : 1172261-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33477
Availability details for
Quantity
  • Availability
  • Add to Cart