ENA159843435 Enamine

5-methyl-N-(3-nitrophenyl)isoxazole-4-carboxamide

  • MDL number MFCD09971657
  • Cas Number : 1049872-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33488
Availability details for
Quantity
  • Availability
  • Add to Cart