ENA159843444 Enamine

4-{3a-methyl-1,5-dioxo-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinazolin-4-yl}butanoic acid

  • MDL number MFCD09971667
  • Cas Number : 1049872-98-1
  • Molecular Weight : 302.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33544
Availability details for
Quantity
  • Availability
  • Add to Cart