ENA159843446 Enamine

methyl 4-[(methylamino)methyl]benzoate hydrochloride

  • MDL number MFCD09971669
  • Cas Number : 160408-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-217
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33547
Availability details for
Quantity
  • Availability
  • Add to Cart