ENA159843549 Enamine

methyl 2-chloro-5-[(dimethylamino)sulfonyl]benzoate

  • MDL number MFCD09971348
  • Cas Number : 1031130-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29842
Availability details for
Quantity
  • Availability
  • Add to Cart