ENA159843553 Enamine

4-(4-methyl-4H-1,2,4-triazol-3-yl)piperidine

  • MDL number MFCD09055373
  • Cas Number : 297172-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31837

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31837
Availability details for
Quantity
  • Availability
  • Add to Cart