ENA159843563 Enamine

5-ethyl-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971570
  • Cas Number : 1152880-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31854

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31854
Availability details for
Quantity
  • Availability
  • Add to Cart