ENA159843573 Enamine

[(adamantan-1-yl)methyl](methyl)amine hydrochloride

  • Cas Number : 1773-99-5
  • Molecular Weight : 215.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 324-325
Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15303

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-15303
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart